Skip to Content
Merck
All Photos(1)

Key Documents

SML4014

Sigma-Aldrich

Edrophonium chloride

≥98% (HPLC)

Synonym(s):

Dimethylethyl(3-hydroxyphenyl)ammonium chloride, Edrophone chloride, Ethyl(m-hydroxyphenyl)dimethylammonium chloride, N-Ethyl-3-hydroxy-N,N-dimethyl-benzenaminium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16NO·Cl
CAS Number:
Molecular Weight:
201.69
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, Faint brown to very dark red brown

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

room temp

SMILES string

OC1=CC=CC([N+](CC)(C)C)=C1.[Cl-]

InChI

1S/C10H15NO.ClH/c1-4-11(2,3)9-6-5-7-10(12)8-9;/h5-8H,4H2,1-3H3;1H

InChI key

BXKDSDJJOVIHMX-UHFFFAOYSA-N

Biochem/physiol Actions

Bioavailable, acetylcholinesterase inhibitor with rapid onset and short duration of action.



Edrophonium chloride is a fast-acting, reversible and competitive inhibitor of acetylcholinesterase (AChE; Ki = 0.2, 0.2, and 0.4 μM in human red blood cells, purified calf forebrain, and octopus brain, respectively) with no effect on butyrylcholinesterase (BChE) activity. Edrophonium dispensing increases acetylcholine (ACh) levels in the neuromuscular junction to result in brief improvements in skeletal and muscular strength. Allosterically interacts with AChE-Serine103 and has a rapid onset of action occurring within 1 minute of administration and a short duration of action lasting 10 minutes with a very short half-life of 2–5 min (intravenous). Atropine, a competitive inhibitor of muscarinic receptors, is an ideal antidote for edrophonium.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Edrophonium
Abdullah Naji
StatPearls [Internet] (2024)
Joakim Holger Waage Brinch et al.
Acta anaesthesiologica Scandinavica, 63(5), 564-575 (2018-12-15)
Mivacurium is a short-acting non-depolarizing muscle relaxant, which is hydrolyzed by butyrylcholinesterase. The neuromuscular block (NMB) can be antagonized with cholinesterase inhibitors (CHEI), but the short duration of action of mivacurium questions the need. This systematic review evaluated if the
M L Andersson et al.
Anaesthesia, 74(4), 518-528 (2019-01-03)
Butyrylcholinesterase deficiency prolongs the effects of the drugs it degrades; succinylcholine and mivacurium. Existing literature on butyrylcholinesterase deficiency is dominated by genetic and biochemical studies. We searched MEDLINE, Embase, Web of Science and Biosis to systematically review the causes and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service