Skip to Content
Merck
All Photos(1)

Key Documents

SML3807

Sigma-Aldrich

BAY 1816032

≥98% (HPLC)

Synonym(s):

2-(3,5-Difluoro-4-((9-(5-methoxy-4-(3-methoxy-pyridin-4-ylamino)-pyrimidin-2-yl)-7,8-diaza-bicyclo[4.3.0]nona-1,3,5,8-tetraen-7-yl)-methyl)-phenoxy)-ethanol, 2-[3,5-Difluoro-4-[[3-[5-methoxy-4-[(3-methoxy-4-pyridinyl)amino]-2-pyrimidinyl]-1H-indazol-1-yl]methyl]phenoxy]ethanol, 2-{3,5-Difluoro-4-[(3-{5-methoxy-4-[(3-methoxypyridin-4-yl)amino]pyrimidin-2-yl}-1H-indazol-1-yl)methyl]phenoxy}ethanol, BAY-1816032, BAY1816032

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H24F2N6O4
CAS Number:
Molecular Weight:
534.51
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

Biochem/physiol Actions

BAY 1816032 is an orally active, ATP-competitive (Ki = 1.2 μM), highly potent and selective BUB1 inhibitor (IC50 = 6.1 nM with 10 μM ATP; affinity Kd = 3.3 nM/BUB1 vs 57 nM/STK10, 850 nM/DMPK2, 2.3 μM/DDR1 and >2.3 μM toward more than 390 other kinases) that effectively targets BUB1-dependent cellular signaling (H2A-pThr120 IC50 = 29 nM post 1h treatment of nocodazole-arrested HeLa cells) and tumor cell proliferation in vitro (IC50 range from 0.5 to 5.8 μM in 43 human and mouse cancer cultures) and in human triple-negative breast xenograft models in vivo (25-50 mg/kg twice daily p.o.; SUM-149 and MDA-MB-436).
Orally active, ATP-competitive, highly potent and selective BUB1 inhibitor with anti-proliferation efficacy against cancer cells in cultures and in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibition of BUB1 Kinase by BAY 1816032 Sensitizes Tumor Cells toward Taxanes, ATR, and PARP Inhibitors In Vitro and In Vivo
Clinical Cancer Research, 25(4), 1404-1414 (2019)
Mapping Proximity Associations of Core Spindle Assembly Checkpoint Proteins.
Journal of Proteome Research, 20(7), 3414-3427 (2021)
Inhibition of BUB1 suppresses tumorigenesis of osteosarcoma via blocking of PI3K/Akt and ERK pathways
Journal of Cellular and Molecular Medicine, 25(17), 8442-8453 (2021)
Biallelic BUB1 mutations cause microcephaly, developmental delay, and variable effects on cohesion and chromosome segregation.
Science Advances, 8(3), eabk0114-eabk0114 (2022)
Miao Zhang et al.
The EMBO journal, 39(3), e101863-e101863 (2019-11-27)
Chromosome segregation in mitosis requires the removal of catenation between sister chromatids. Timely decatenation of sister DNAs at mitotic centromeres by topoisomerase IIα (TOP2A) is crucial to maintain genomic stability. The chromatin factors that recruit TOP2A to centromeres during mitosis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service