Skip to Content
Merck
All Photos(1)

Documents

SML3486

Sigma-Aldrich

PFI-7N hydrochloride

≥98% (HPLC)

Synonym(s):

N-((1s,4S)-4-(1H-benzo[d]imidazol-2-yl)cyclohexyl)-2-((R)-1-phenylethylamino)acetamide hydrochloride, PFI-7 inactive control hydrochloride, PFI-7 negative control hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H28N4O · xHCl
Molecular Weight:
376.49 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(CN[C@H](C)C1=CC=CC=C1)N[C@@H]2CC[C@@H](CC2)C3=NC4=C(N3)C=CC=C4

Biochem/physiol Actions

PFI-7N is an inactive analog of and negative control for the high affinity E3 ligase GID4 probe PFI-7 (KD = 5 μM/PFI-7N vs. 0.08 μM/PFI-7 by SPR).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service