Skip to Content
Merck
All Photos(1)

Documents

SML3420

Sigma-Aldrich

KY-226

≥97% (HPLC)

Synonym(s):

4-((Biphenyl-4-ylmethylthio)methyl)-N-(hexylsulfonyl)benzamide, 4-(Biphenyl-4-ylmethylsulfanylmethyl)-N-(hexane-1-sulfonyl)benzoylamide, 4-[[([1,1′-Biphenyl]-4-ylmethyl)thio]methyl]-N-(hexylsulfonyl)benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H31NO3S2
CAS Number:
Molecular Weight:
481.67
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NS(=O)(CCCCCC)=O)C1=CC=C(CSCC2=CC=C(C3=CC=CC=C3)C=C2)C=C1

Biochem/physiol Actions

KY-226 is an orally bioavailable, potent and selective allosteric inhibitor of protein tyrosine phosphatase 1B (PTP1B) that does not function as a PPARg agonist. KY-226 exhibits anti-diabetic, anti-obesity effects by enhancement of insulin and leptin signaling in mice. It appears that KY-226 exerts neuroprotective effects through restoration of phosphorylation of Akt and FoxO1. Thus, it protects BBB integrity by restoration of tight junction (TJ) proteins in mice.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service