Skip to Content
Merck
All Photos(1)

Key Documents

SML3257

Sigma-Aldrich

Demethyleneberberine

≥98% (HPLC)

Synonym(s):

5,​6-​Dihydro-​2,​3-​dihydroxy-​9,​10-​dimethoxy-dibenzo[a,​g]​quinolizinium, 7,8,13,13a-Tetradehydro-2,3-dihydroxy-9,10-dimethoxy-berbinium, Demethylene-berberine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18NO4
CAS Number:
Molecular Weight:
324.35
UNSPSC Code:
12352210
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, White to very dark grey

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

[n+]21c(cc4c(c2)c(c(cc4)OC)OC)c3c(cc(c(c3)O)O)CC1

InChI

1S/C19H17NO4/c1-23-18-4-3-11-7-15-13-9-17(22)16(21)8-12(13)5-6-20(15)10-14(11)19(18)24-2/h3-4,7-10,22H,5-6H2,1-2H3/p+1

InChI key

HVTCKKMWZDDWOY-UHFFFAOYSA-O

Biochem/physiol Actions

Demethyleneberberine, a berberine metabolite, is a mitochondria-targeted antioxidant isolated from Cortex Phellodendri chinensis that exhibits multiple pharmacological activities including anti-microbial, anti-inflammatory, anti-diarrhea and anti-cancer. Demethyleneberberine alleviates ethanol-dependent oxidative stress by suppression of cytochrome CYP2E1 (P450 2E1), hypoxia inducible factor α (HIF-1α) and inducible nitric oxide synthase (iNOS). It appears that demethyleneberberine attenuates non-alcoholic fatty liver disease (NAFLD) through activation of AMPK.
Mitochondrially targeted antioxidant

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi-Ting Zhang et al.
Acta pharmacologica Sinica, 40(1), 133-142 (2018-11-18)
Berberine, berberrubine, thalifendine, demethyleneberberine, jatrorrhizine, and columbamine are six natural protoberberine alkaloid (PA) compounds that display extensive pharmacological properties and share the same protoberberine molecular skeleton with only slight substitution differences. The oral delivery of most PAs is hindered by
Xiaoyan Qiang et al.
Biochemical and biophysical research communications, 472(4), 603-609 (2016-03-13)
Non-alcoholic fatty liver disease (NAFLD) has reached an epidemic level globally, which is recognized to form non-alcoholic steatohepatitis (NASH) by the "two-hit" model, including oxidative stress and inflammation. AMP-activated protein kinase (AMPK) has long been regarded as a key regulator
Pengcheng Zhang et al.
The Journal of pharmacology and experimental therapeutics, 352(1), 139-147 (2014-11-02)
Excessive alcohol consumption induces oxidative stress and lipid accumulation in the liver. Mitochondria have long been recognized as the key target for alcoholic liver disease (ALD). Recently, the artificial mitochondria-targeted antioxidant MitoQ has been used to treat ALD effectively in

Questions

  1. Hi, I would like to ask if your product Demethyleneberberine (CAS: 25459-91-0) has anionic binding? If so, what anion is it? Is it chloride? Many thanks!

    1 answer
    1. This product is provided in its free-amine form without a counter ion, e.g., an anion bound to it.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service