Skip to Content
Merck
All Photos(1)

Key Documents

SML3111

Sigma-Aldrich

ETP-47799

≥98% (HPLC)

Synonym(s):

CDK8/19i-47799, CDK8/19i-CNIO-47799, ETP 47799, ETP47799, Lck-IN-1, N-Propyl-3-(4-pyridinyl)imidazo[1,2-b]pyridazin-6-amine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
CHF 101.00
25 MG
CHF 378.00

CHF 101.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
CHF 101.00
25 MG
CHF 378.00

About This Item

Empirical Formula (Hill Notation):
C14H15N5
CAS Number:
Molecular Weight:
253.30
UNSPSC Code:
12352200
NACRES:
NA.77

CHF 101.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CCCNC1=NN2C(C=C1)=NC=C2C3=CC=NC=C3

InChI key

XTMROGQLIVGYKF-UHFFFAOYSA-N

Biochem/physiol Actions

ETP-47799 is a potent and CDK8-/CDK19-selective cyclin-dependent kinase inhibtor (CDK8/19 IC50 = 2.9/14.1 nM; CDK1/5/9 IC50 = 275/239/124 nM; CDK2/4/6/7 IC50 >1 μM; GSK3β IC50 = 3.14 μM) with additional potency against PDGFRα (IC50 = 3.5 nM). ETP-47799 ( 0.4 μM or 1.1 μM) induces and stabilizes the naive state of murine and human pluripotent stem cells (PSCs) with better efficiency than the commonly used MEK/GSK3 inhibitor cocktail (1μM PD0325901, 1.5μM CHIR 99021).
Potent and CDK8-/CDK19-selective cyclin-dependent kinase inhibtor that induces and stabilizes the naive state of murine and human pluripotent stem cells (PSCs).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service