Skip to Content
Merck
All Photos(1)

Key Documents

SML3093

Sigma-Aldrich

2′-MeCCPA

≥98% (HPLC)

Synonym(s):

2′-Me-CCPA, 2-Chloro-2′-C-methyl-N6-cyclopentyladenosine, 2-Chloro-N-cyclopentyl-2′-C-methyladenosine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
CHF 113.00
25 MG
CHF 453.00

CHF 113.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
CHF 113.00
25 MG
CHF 453.00

About This Item

Empirical Formula (Hill Notation):
C16H22ClN5O4
CAS Number:
Molecular Weight:
383.83
UNSPSC Code:
12352200
NACRES:
NA.77

CHF 113.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

Clc1nc2[n](cnc2c(n1)NC4CCCC4)C3OC(C(C3(O)C)O)CO

InChI

1S/C16H22ClN5O4/c1-16(25)11(24)9(6-23)26-14(16)22-7-18-10-12(19-8-4-2-3-5-8)20-15(17)21-13(10)22/h7-9,11,14,23-25H,2-6H2,1H3,(H,19,20,21)

InChI key

MMPAUXMIDJWGFO-UHFFFAOYSA-N

Biochem/physiol Actions

2′-MeCCPA is a potent and A1-selective adenosine receptor agonist (Ki = 1.8 nM/bovine A1, 3.9 μM/bovine A2a, 5.0 μM/rat A3). 2′-MeCCPA inhibits 100 μM forskolin-induced adenylate cyclase activity in rat cortical membrane preparations (IC50 = 13.1 nM; ECmax = 15.1% inhibition) without antagonist potency against A1 agonist CHA (N6-cyclohexyladenosine).
Potent and A1-selective adenosine receptor agonist with no antagonist potency.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service