Skip to Content
Merck
All Photos(1)

Key Documents

SML3001

Sigma-Aldrich

Phleomycin ready made solution

0.2 μm filtered, 20mg/mL in 20mM HEPES

Synonym(s):

Phleomycin ready made solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
CHF 86.20

CHF 86.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
CHF 86.20

About This Item

Empirical Formula (Hill Notation):
C55H86N20O21S2
Molecular Weight:
1427.52
UNSPSC Code:
41121800
NACRES:
NA.77

CHF 86.20


Check Cart for Availability

Request a Bulk Order

Quality Level

sterility

0.2 μm filtered

form

liquid

storage condition

protect from light

storage temp.

−20°C

SMILES string

N=C(NCCCCNC(C1=CSC(C2CSC(CCNC(C(NC(C(C(C(NC(C([C@H](O[C@H]3[C@@H](O[C@@H]4O[C@@H](CO)[C@H](O)[C@H](OC(N)=O)C4O)[C@@H](O)[C@H](O)[C@H](CO)O3)C5=CN=CN5)NC(C6=C(C)C(N)=NC(C(NCC(C(N)=O)N)CC(N)=O)=N6)=O)=O)C)O)C)=O)C(O)C)=O)=N2)=N1)=O)N

General description

Phleomycin is a glycopeptide antibiotic produced by Streptomyces verticillus. It is a member of Bleomycin family with wide spectrum antibiotic activity in both prokaryotic and eukaryotic cells. In resemblance to Bleomycin activity, Phleomycin is also an anti-cancer agent that causes DNA cleavage which leads to cell death. DNA cleavage by Phleomycin is initiated by metal ions, causing reactive oxidative species (ROS) which induces oxidative damage.Therefore, Phleomycin is used as a DNA damage inducer to test DNA double strand repair. Additionally, DNA repair is tested by comet assay to measure DNA breaks after Phleomycin treatment. Phleomycin is also commonly used as a selective agent by utilizing ble gene to induce cell resistance. The ble gene encodes BLE protein which forms a 1:1 complex with phleomycin, decreasing its toxicity.

Biochem/physiol Actions

Antibiotic, Selection agent, Anticancer agent

Other Notes

  • Phleomycin ready made solution is provided in a neutral buffer (20mM HEPES, pH∼7). Due to its sensitivity to acidic or alkaline medium neutral environment should be kept.
  • Phleomycin ready made solution is light sensitive and should be kept protected from light.
  • Avoid repeated freeze thaw cycle of this product. Phleomycin ready made solution should be aliquoted upon receipt.
  • The recommended concentration for phleomycin-induced DNA damage applications is 2.5-100μg/mL7-12, therefore stock solution (20mg/mL) can be diluted by 200-8000 folds.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service