Skip to Content
Merck
All Photos(1)

Key Documents

SML2602

Sigma-Aldrich

CW3388

≥98% (HPLC)

Synonym(s):

1-([1,1′-Biphenyl]-4-ylmethyl)octahydro-azocine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
CHF 88.00
25 MG
CHF 342.00

CHF 88.00


Estimated to ship on17 May 2025



Select a Size

Change View
5 MG
CHF 88.00
25 MG
CHF 342.00

About This Item

Empirical Formula (Hill Notation):
C20H25N
CAS Number:
Molecular Weight:
279.42
UNSPSC Code:
12352200
NACRES:
NA.77

CHF 88.00


Estimated to ship on17 May 2025


Assay

≥98% (HPLC)

form

oil

color

colorless to yellow

storage temp.

2-8°C

SMILES string

N3(CCCCCCC3)Cc1ccc(cc1)c2ccccc2

InChI

1S/C20H25N/c1-2-7-15-21(16-8-3-1)17-18-11-13-20(14-12-18)19-9-5-4-6-10-19/h4-6,9-14H,1-3,7-8,15-17H2

InChI key

BTDKOFMWYIXWST-UHFFFAOYSA-N

Biochem/physiol Actions

CW3388 is a cell active and highly potent enhancer of oligodendrocyte formation. CW3388 potently inhibits the cholesterol biosynthesis enzyme Δ8,7-sterol isomerase (EBP). It promotes myelination in vitro.
cell active and highly potent enhancer of oligodendrocyte formation; inhibitor of Δ8,7-sterol isomerase (EBP)

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dharmaraja Allimuthu et al.
Cell chemical biology, 26(4), 593-599 (2019-02-19)
Small molecules that promote oligodendrocyte formation have been identified in "drug repurposing" screens to nominate candidate therapeutics for diseases in which myelin is lost, including multiple sclerosis. We recently reported that many such molecules enhance oligodendrocyte formation not by their

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service