Skip to Content
Merck
All Photos(1)

Key Documents

SML2535

Sigma-Aldrich

MRK-740

≥98% (HPLC)

Synonym(s):

4-(3-(3,5-Dimethoxy-phenyl)-1,2,4-oxadiazol-5-yl)-1-methyl-9-(2-methyl-pyridin-4-yl)-1,4,9-triaza-spiro[5.5]undecane

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
CHF 118.00
25 MG
CHF 473.00

CHF 118.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
CHF 118.00
25 MG
CHF 473.00

About This Item

Empirical Formula (Hill Notation):
C25H32N6O3
CAS Number:
Molecular Weight:
464.56
UNSPSC Code:
51111800
NACRES:
NA.77

CHF 118.00


Estimated to ship on25 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CN1CCN(C2=NC(C3=CC(OC)=CC(OC)=C3)=NO2)CC14CCN(C5=CC(C)=NC=C5)CC4

InChI

1S/C25H32N6O3/c1-18-13-20(5-8-26-18)30-9-6-25(7-10-30)17-31(12-11-29(25)2)24-27-23(28-34-24)19-14-21(32-3)16-22(15-19)33-4/h5,8,13-16H,6-7,9-12,17H2,1-4H3

InChI key

NZYTZRHHBAJPKN-UHFFFAOYSA-N

Biochem/physiol Actions

MRK-740 is a cell penetrant, potent and selective inhibitor of PRDM9 that inhibits methylation of H3K4 in cells.
cell penetrant, potent and selective inhibitor of PRDM9

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abdellah Allali-Hassani et al.
Nature communications, 10(1), 5759-5759 (2019-12-19)
PRDM9 is a PR domain containing protein which trimethylates histone 3 on lysine 4 and 36. Its normal expression is restricted to germ cells and attenuation of its activity results in altered meiotic gene transcription, impairment of double-stranded breaks and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service