Skip to Content
Merck
All Photos(1)

Key Documents

SML2224

Sigma-Aldrich

MS436

≥98% (HPLC)

Synonym(s):

(E)-4-[2-(2-Amino-4-hydroxy-5-methylphenyl)diazenyl]-N-2-pyridinylbenzenesulfonamide, 4-[(1E)-2-(2-Amino-4-hydroxy-5-methylphenyl)diazenyl]-N-2-pyridinylbenzenesulfonamide, MS 436, MS-436

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
CHF 68.50
25 MG
CHF 277.00

CHF 68.50


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
CHF 68.50
25 MG
CHF 277.00

About This Item

Empirical Formula (Hill Notation):
C18H17N5O3S
CAS Number:
Molecular Weight:
383.42
MDL number:
UNSPSC Code:
12352200

CHF 68.50


Estimated to ship on21 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

faint brown to dark red

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=S(C(C=C1)=CC=C1/N=N/C2=C(N)C=C(O)C(C)=C2)(NC3=NC=CC=C3)=O

InChI

1S/C18H17N5O3S/c1-12-10-16(15(19)11-17(12)24)22-21-13-5-7-14(8-6-13)27(25,26)23-18-4-2-3-9-20-18/h2-11,24H,19H2,1H3,(H,20,23)

InChI key

DZTGIRNXWSZBIM-UHFFFAOYSA-N

Biochem/physiol Actions

BRD4 inhibitor with selective affinity toward the first bromodomain (BrD1) over BRD4 BrD2, CBP BrD, PCAF BrD.
MS436 is a diazobenzene-based potent BRD4 inhibitor with selective affinity toward the first bromodomain (BrD) of BRD4 (BRD4 BrD1 Ki <85 nM vs. BRD4 BrD2 Ki = 340 nM; IC50 = 460 nM/BRD4 BrD1 vs. 1.29 μM/BRD4 BrD2, 4.95 μM/CBP BrD, 20.29 μM/PCAF BrD). MS436 is a useful tool for probing BRD4-dependent cellular functions, including NF-κB-mediated proinflammatory response in RAW264.7 murine macrophages (IC50 in μM = 4.9/3.8 against LPS-induced IL-6/NO production), human and murine embryonic stem cell (ESC) pluripotency maintenance (10 μM), telomere elongation in mTERT/mTR-overexpressing murine fibroblasts (5 μM), and bioenergetic deficiency caused by mitochondrial complex I (CI)-mutation in human cybrid cells (0.9 μM).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steven Wang et al.
Nucleic acids research, 45(14), 8403-8410 (2017-09-01)
Cancer cells maintain telomere length equilibrium to avoid senescence and apoptosis induced by short telomeres, which trigger the DNA damage response. Limiting the potential for telomere maintenance in cancer cells has been long been proposed as a therapeutic target. Using
Raffaella Di Micco et al.
Cell reports, 9(1), 234-247 (2014-09-30)
Transcription factors and chromatin-remodeling complexes are key determinants of embryonic stem cell (ESC) identity. Here, we demonstrate that BRD4, a member of the bromodomain and extraterminal domain (BET) family of epigenetic readers, regulates the self-renewal ability and pluripotency of ESCs.
Rosalia Fernandez-Alonso et al.
EMBO reports, 18(7), 1108-1122 (2017-06-08)
Pluripotent stem cells (PSCs) hold great clinical potential, as they possess the capacity to differentiate into fully specialised tissues such as pancreas, liver, neurons and cardiac muscle. However, the molecular mechanisms that coordinate pluripotent exit with lineage specification remain poorly
Gurkan Mollaoglu et al.
Cancer cell, 31(2), 270-285 (2017-01-17)
Loss of the tumor suppressors RB1 and TP53 and MYC amplification are frequent oncogenic events in small cell lung cancer (SCLC). We show that Myc expression cooperates with Rb1 and Trp53 loss in the mouse lung to promote aggressive, highly
Joeva J Barrow et al.
Molecular cell, 64(1), 163-175 (2016-09-27)
Mitochondrial diseases comprise a heterogeneous group of genetically inherited disorders that cause failures in energetic and metabolic function. Boosting residual oxidative phosphorylation (OXPHOS) activity can partially correct these failures. Herein, using a high-throughput chemical screen, we identified the bromodomain inhibitor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service