Skip to Content
Merck
All Photos(1)

Key Documents

SML2193

Sigma-Aldrich

Altenusin

≥98% (HPLC)

Synonym(s):

3,4′,5′-Trihydroxy-5-methoxy-2′-methyl[1,1′-biphenyl]-2-carboxylic acid, Alutenusin

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
CHF 161.00

CHF 161.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
CHF 161.00

About This Item

Empirical Formula (Hill Notation):
C15H14O6
CAS Number:
Molecular Weight:
290.27
UNSPSC Code:
12352200
NACRES:
NA.77

CHF 161.00


Check Cart for Availability

Request a Bulk Order

biological source

Penicillium sp.

Assay

≥98% (HPLC)

form

powder

color

yellow to orange

storage temp.

−20°C

SMILES string

OC1=CC(C2=CC(OC)=CC(O)=C2C(O)=O)=C(C)C=C1O

InChI

1S/C15H14O6/c1-8-5-13(16)14(17)7-11(8)12-6-9(20-2)3-4-10(12)15(18)21-19/h3-7,16-17,19H,1-2H3

InChI key

GWGIZSANXIQQQR-UHFFFAOYSA-N

Biochem/physiol Actions

Altenusin, a non-steroidal fungal metabolite, is a potent and selective agonist of Farnesoid X receptor (FXR) that protects mice from high-fat diet induced obesity. Also it decreases blood glucose level, serum insulin level, and reduces lipid droplets accumulation. Altenusin inhibits trypanothione reductase from Trypanosoma cruzi.
potent and selective agonist of Farnesoid X receptor (FXR)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sook Wern Chua et al.
ACS chemical neuroscience, 8(4), 743-751 (2017-01-10)
In Alzheimer's disease, the microtubule-associated protein tau forms intracellular neurofibrillary tangles (NFTs). A critical step in the formation of NFTs is the conversion of soluble tau into insoluble filaments. Accordingly, a current therapeutic strategy in clinical trials is aimed at
Betania Barros Cota et al.
FEMS microbiology letters, 285(2), 177-182 (2008-06-19)
Parasitic protozoan species belonging to the genera Trypanosoma and Leishmania are the etiological agents of several diseases in tropical areas of the world, for which there is an urgent need for effective and affordable treatment. In this regard, we are
Zhihui Zheng et al.
Molecular pharmacology, 92(4), 425-436 (2017-07-26)
Nonalcoholic fatty liver disease (NAFLD) is a prevalent chronic liver disease. The incidence of NAFLD has increased steadily due to its close association with the global epidemic of obesity and type 2 diabetes. However, there is no effective pharmacological therapy
Christian M Garrido et al.
Scientific reports, 10(1), 9120-9120 (2020-06-06)
K-Ras must interact primarily with the plasma membrane (PM) for its biological activity. Therefore, disrupting K-Ras PM interaction is a tractable approach to block oncogenic K-Ras activity. Here, we found that avicin G, a family of natural plant-derived triterpenoid saponins
Jian Shi et al.
Cell reports, 33(1), 108225-108225 (2020-10-08)
Endogenous PIEZO1 channels of native endothelium lack the hallmark inactivation often seen when these channels are overexpressed in cell lines. Because prior work showed that the force of shear stress activates sphingomyelinase in endothelium, we considered if sphingomyelinase is relevant

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service