Skip to Content
Merck
All Photos(1)

Key Documents

SML2095

Sigma-Aldrich

Nalfurafine hydrochloride

≥98% (HPLC)

Synonym(s):

(2E)-N-[(5α,6β)-17-(cyclopropylmethyl)- 3,14-dihydroxy- 4,5-epoxymorphinan- 6-yl]- 3-(3-furyl)-N-methylacrylamide hydrochloride, AC-820 HCl, TRK-820 HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H32N2O5 · HCl
CAS Number:
Molecular Weight:
513.03
MDL number:
UNSPSC Code:
12352202

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C28H32N2O5.ClH/c1-29(23(32)7-4-18-9-13-34-16-18)20-8-10-28(33)22-14-19-5-6-21(31)25-24(19)27(28,26(20)35-25)11-12-30(22)15-17-2-3-17;/h4-7,9,13,16-17,20,22,26,31,33H,2-3,8,10-12,14-15H2,1H3;1H/b7-4+;/t20-,22-,26+,27+,28-;/m1./s1

InChI key

DJSFYNINGIMKAG-FQJQBBMWSA-N

Biochem/physiol Actions

Nalfurafine (TRK-820) is a selective κ-opioid (KOR) agonist with antipruritic activity.
Potent and selective kappa opioid receptor agonist

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nalfurafine hydrochloride, a selective κ opioid receptor agonist, has no reinforcing effect on intravenous self-administration in rhesus monkeys.
Xiulu Ruan et al.
Journal of pharmacological sciences, 132(1), 113-114 (2016-07-28)
H Nagase et al.
Chemical & pharmaceutical bulletin, 46(2), 366-369 (1998-03-21)
A new type of kappa-agonist, 17-cyclopropylmethyl-3, 14 beta-dihydroxy-4,5 alpha-epoxy-6 beta-[N-methyl-trans-3-(3-furyl) acrylamido]morphinan hydrochloride (1, TRK-820), was discovered by a new working hypothesis. The "message-address concept" for opioid antagonists and the "accessory site" for general antagonists were applied to design TRK-820. A
Selena S Schattauer et al.
Cellular signalling, 32, 59-65 (2017-01-16)
Nalfurafine is a moderately selective kappa opioid receptor (KOR) analgesic with low incidence of dysphoric side effects in clinical development for the treatment of uremic pruritis. The basis for its reduced dysphoric effect compared to other KOR agonists is not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service