Skip to Content
Merck
All Photos(1)

Key Documents

SML1984

Sigma-Aldrich

GSK9311

≥98% (HPLC)

Synonym(s):

(R)-N-(1,3-Dimethyl-6-(2-methylpiperazin-1-yl)-2-oxo-2,3-dihydro-1H-benzo[d]imidazol-5-yl)-N-ethyl-2-methoxybenzamide, N-[2,3-Dihydro-1,3-dimethyl-6-[(2R)-2-methyl-1-piperazinyl]-2-oxo-1H-benzimidazol-5-yl]-N-ethyl-2-methoxybenzamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
CHF 90.10

CHF 90.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
CHF 90.10

About This Item

Empirical Formula (Hill Notation):
C24H31N5O3
CAS Number:
Molecular Weight:
437.53
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

CHF 90.10


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D 90 to 100°, c = 0.3 in methanol

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

O=C1N(C)C2=CC(N3CCNC[C@H]3C)=C(N(CC)C(C4=C(OC)C=CC=C4)=O)C=C2N1C

InChI key

WFXIHQFRQPGCCR-MRXNPFEDSA-N

Biochem/physiol Actions

GSK9311 is a negative control for the SGC epigenetic probe GSK6853. GSK9311 exhibits 125- and 185-fold reduced potency than its structural analog GSK6853 toward BRPF1 in cell-free and cell-based assays (pIC50 = 6.0/GSK9311 vs. 8.1/GSK6853 in ligand competition binding assay by TR-FRET; IC50 = 3.7 μM/GSK9311 vs. 20 nM/GSK6853 against NanoLuc-BRPF1 bromodomain interaction with Histone H3.3-HaloTag in HEK293 cells), and therefore serves as a good inactive control compound in BRPF1 inhibition studies employing GSK6853. For characterization details of the active probe, GSK6853, please visit the GSK6853 probe summary on the Structural Genomics Consortium (SGC) website.

GSK6853, the active probe, is available from Sigma. To learn more about and purchase GSK6853, click here.

To learn about other SGC chemical probes for epigenetic targets, visit sigma.com/sgc
Less potent control compound for GSK6853 in BRPF1 inhibition studies.

Features and Benefits

GSK9311 is the negative control for GSK6853, an epigenetic chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC epigenetic probes, visit sigma.com/SGC.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paul Bamborough et al.
ACS medicinal chemistry letters, 7(6), 552-557 (2016-06-22)
The BRPF (Bromodomain and PHD Finger-containing) protein family are important scaffolding proteins for assembly of MYST histone acetyltransferase complexes. A selective benzimidazolone BRPF1 inhibitor showing micromolar activity in a cellular target engagement assay was recently described. Herein, we report the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service