Skip to Content
Merck
All Photos(1)

Key Documents

SML1913

Sigma-Aldrich

ASB14780

≥98% (HPLC)

Synonym(s):

3-(3-Phenethyl-1-(4-phenoxyphenyl)-1H-indol-5-yl)propanoic acid, 2-amino-2-(hydroxymethyl)-1,3-propanediol, 3-[1-(4-Phenoxyphenyl)-3-(2-phenylethyl)-1H-indol-5-yl]propanoic acid, 2-amino-2-(hydroxymethyl)-1,3-propanediol

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
CHF 125.00
25 MG
CHF 506.00

CHF 125.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
CHF 125.00
25 MG
CHF 506.00

About This Item

Empirical Formula (Hill Notation):
C31H27NO3 · C4H11NO3
CAS Number:
Molecular Weight:
582.69
UNSPSC Code:
12352200
NACRES:
NA.77

CHF 125.00


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(O)CCC1=CC2=C(N(C3=CC=C(OC4=CC=CC=C4)C=C3)C=C2CCC5=CC=CC=C5)C=C1.NC(CO)(CO)CO

InChI

1S/C31H27NO3.C4H11NO3/c33-31(34)20-13-24-12-19-30-29(21-24)25(14-11-23-7-3-1-4-8-23)22-32(30)26-15-17-28(18-16-26)35-27-9-5-2-6-10-27;5-4(1-6,2-7)3-8/h1-10,12,15-19,21-22H,11,13-14,20H2,(H,33,34);6-8H,1-3,5H2

InChI key

MBPXGBINIINSEU-UHFFFAOYSA-N

Biochem/physiol Actions

ASB14780 can be a potential therapeutic for the treatment of asthma and other pulmonary diseases.[1] In addition, its inhibitory function on group IVA phospholipase A2 (IVA-PLA2) qualifies this drug to be a potential candidate for the treatment of non-alcoholic fatty liver diseases.[2]
ASB14780 is an indole-based selective and potent cytosolic phospholipase A2α inhibitor (IC50 = 20 nM/cPLA2α and >10 μM/sPLA2α). ASB14780 inhibits cPLA2α-dependent inflammatory responses both in cultures and in animals in vivo, including LPS-induced PGE2 production (IC50 = 0.5 μM; mouse peritoneal exudate cells), A23187-induced TXB2 production (IC50 = 0.54 and 0.64 μM using guinea pig and human whole blood, respectively), TPA-induced ear edema (50 mg/kg, p.o.; mice), OVA-induced asthma (5 - 20 mg/kg/d, p.o.; guinea pig), CCl4-induced hepatic fibrosis and high-fat cholesterol diet-induced fatty liver (0.1-0.3 g/kg/d, p.o.; mouse). SB14780 bioavailability is also demonstrated in dog and monkey, albeit at a lower value (F = 34.3%/dog and 30.9%/monkey vs. 89.6%/mouse).
ASB14780 is an indole-based selective and potent cytosolic phospholipase A2α inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shiho Kanai et al.
The Journal of pharmacology and experimental therapeutics, 356(3), 604-614 (2015-12-25)
We have previously shown that high-fat cholesterol diet (HFCD)-induced fatty liver and carbon tetrachloride (CCl4)-induced hepatic fibrosis are reduced in mice deficient in group IVA phospholipase A2 (IVA-PLA2), which plays a role in inflammation. We herein demonstrate the beneficial effects
Toshiyuki Tomoo et al.
Journal of medicinal chemistry, 57(17), 7244-7262 (2014-08-08)
This article describes the design, synthesis, and biological evaluation of new indole-based cytosolic phospholipase A2α (cPLA2α, a group IVA phospholipase A2) inhibitors. A screening-hit compound from our library, (E)-3-{4-[(4-chlorophenyl)thio]-3-nitrophenyl}acrylic acid (5), was used to design a class of 3-(1-aryl-1H-indol-5-yl)propanoic acids

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service