Skip to Content
Merck
All Photos(1)

Documents

SML1855

Sigma-Aldrich

AZD5438

≥97% (HPLC)

Synonym(s):

4-[2-Methyl-1-(1-methylethyl)-1H-imidazol-5-yl]-N-[4-(methylsulfonyl)phenyl]-2-pyrimidinamine, AZD 5438, AZD-5438

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21N5O2S
CAS Number:
Molecular Weight:
371.46
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

powder

color

yellow to orange

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C18H21N5O2S/c1-12(2)23-13(3)20-11-17(23)16-9-10-19-18(22-16)21-14-5-7-15(8-6-14)26(4,24)25/h5-12H,1-4H3,(H,19,21,22)

InChI key

WJRRGYBTGDJBFX-UHFFFAOYSA-N

Biochem/physiol Actions

AZD5438 is an orally active, potent and reversible inhibitor against cyclin-dependent kinases (IC50 = 6 nM/Cyc E-CDK2, 14 nM/p25-CDK5, 16 nM/Cyc B1-CDK1, 20 nM/Cyc T-CDK9, 21 nM/Cyc D3-CDK6, 45 nM/Cyc A-CDK2, 449 nM/Cyc D1-CDK4, 821 nM/Cyc H-CDK7; [ATP] = Km) and glycogen synthase kinase GSK-3β≤ (IC50 = 17 nM) without significant inhibitory potency toward 30 other kinases (≤75% inhibition at 10 μM). AZD5438 effectively inhibits cellular CDK substrates phosphorylation and displays antiproliferation activity against a broad spectrum of human cancer cultures (IC50 from 0.2 μM/MCF to 1.7 μM/ARH-77) by inducing cell cycle arrest at G2-M, S, and G1. Oral administration (50 mg/kg/12 h or 75 mg/kg/day) is efficacious against human tumor xenograft growth in mice in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kate F Byth et al.
Molecular cancer therapeutics, 8(7), 1856-1866 (2009-06-11)
Deregulation of the cell cycle has long been recognized as an essential driver of tumorigenesis, and agents that selectively target key cell cycle components continue to hold promise as potential therapeutics. We have developed AZD5438, a 4-(1-isopropyl-2-methylimidazol-5-yl)-2-(4-methylsulphonylanilino) pyrimidine, as a
Nozomi Tomimatsu et al.
Nature communications, 5, 3561-3561 (2014-04-08)
Resection of DNA double-strand breaks (DSBs) is a pivotal step during which the choice between NHEJ and HR DNA repair pathways is made. Although CDKs are known to control initiation of resection, their role in regulating long-range resection remains elusive.
Camille D A Terfve et al.
Nature communications, 6, 8033-8033 (2015-09-12)
Mass spectrometry is widely used to probe the proteome and its modifications in an untargeted manner, with unrivalled coverage. Applied to phosphoproteomics, it has tremendous potential to interrogate phospho-signalling and its therapeutic implications. However, this task is complicated by issues

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service