Skip to Content
Merck
All Photos(1)

Key Documents

SML1773

Sigma-Aldrich

TYFAVLM trifluoroacetate salt

≥98% (HPLC)

Synonym(s):

P7, Thr-Tyr-Phe-Ala-Val-Leu-Met trifluoroacetate salt, synthetic P7 peptide agonist trifluoroacetate salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
CHF 184.00

CHF 184.00


Estimated to ship on27 April 2025



Select a Size

Change View
1 MG
CHF 184.00

About This Item

Empirical Formula (Hill Notation):
C41H61N7O10S · xC2HF3O2
Molecular Weight:
844.03 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

CHF 184.00


Estimated to ship on27 April 2025


Quality Level

Assay

≥98% (HPLC)

form

lyophilized powder

storage condition

protect from light

color

white

shipped in

wet ice

storage temp.

−20°C

Biochem/physiol Actions

Potent and selective agonist of GPR56
TYFAVLM/P7, a synthetic peptides comprising adhesion GPR56 β-strand-13 stalk, is a potent and selective agonist of GPR56. TYFAVLM activates GPR56 in cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oladapo E Olaniru et al.
Molecular metabolism, 53, 101285-101285 (2021-07-06)
Members of the adhesion G protein-coupled receptor (aGPCR) subfamily are important actors in metabolic processes, with GPR56 (ADGRG1) emerging as a possible target for type 2 diabetes therapy. GPR56 can be activated by collagen III, its endogenous ligand, and by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service