Skip to Content
Merck
All Photos(1)

Documents

SML1700

Sigma-Aldrich

(S)-ZINC-3573

≥98% (HPLC)

Synonym(s):

(S)-N,N-Dimethyl-1-(5-phenylpyrazolo[1,5-a]pyrimidin-7-yl)-3-pyrrolidinamine, (S)-ZINC7243573

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21N5
CAS Number:
Molecular Weight:
307.39
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CN([C@@H](CC1)CN1C2=CC(C3=CC=CC=C3)=NC4=CC=NN24)C

Application

(S)-ZINC-3573 has been used to investigate the biology of the primate-exclusive MRGPRX2 G protein-coupled receptor (GPCR).

Biochem/physiol Actions

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In silico design of novel probes for the atypical opioid receptor MRGPRX2
Lansu K, et al.
Nature Chemical Biology, 13(5), 529-529 (2017)
In silico design of novel probes for the atypical opioid receptor MRGPRX2
Lansu, K.
Nature Chemical Biology (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service