Skip to Content
Merck
All Photos(1)

Key Documents

SML1677

Sigma-Aldrich

Tarocin A1

≥98% (HPLC)

Synonym(s):

(4S,5R)-5-(3,5-bis(trifluoromethyl)phenyl)-3-(3-(isoquinolin-4-yl)propanoyl)-4-methyloxazolidin-2-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
CHF 97.30
25 MG
CHF 392.00

CHF 97.30


Estimated to ship on27 April 2025


Request a Bulk Order

Select a Size

Change View
5 MG
CHF 97.30
25 MG
CHF 392.00

About This Item

Empirical Formula (Hill Notation):
C24H18F6N2O3
CAS Number:
Molecular Weight:
496.40
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

CHF 97.30


Estimated to ship on27 April 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

C[C@H](N1C(CCC2=C(C=CC=C3)C3=CN=C2)=O)[C@@H](C4=CC(C(F)(F)F)=CC(C(F)(F)F)=C4)OC1=O

InChI

1S/C24H18F6N2O3/c1-13-21(16-8-17(23(25,26)27)10-18(9-16)24(28,29)30)35-22(34)32(13)20(33)7-6-15-12-31-11-14-4-2-3-5-19(14)15/h2-5,8-13,21H,6-7H2,1H3/t13-,21-/m0/s1

InChI key

XWMRKUPNQVITEC-ZSEKCTLFSA-N

Biochem/physiol Actions

Tarocin A1 is an oxazolidinone which inhibits TarO, the first enzyme in teichoic acid synthesis.
Tarocin A1 is an oxazolidinone which inhibits TarO, the first enzyme in teichoic acid synthesis. Tarocin A1 has an IC50 value of 40 nM in a biochemical TarO assay. Tarocin A does not inhibit bacterial growth alone, and exhibits no cytotoxicity against HeLa cells, but it has potent bactericidal syergism with broad spectrum β-lactams, and may restore the clinical efficacy of existing β-lactam antibiotics.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service