Skip to Content
Merck
All Photos(1)

Key Documents

SML1550

Sigma-Aldrich

Zibotentan

≥98% (HPLC)

Synonym(s):

N-(3-methoxy-5-methylpyrazin-2-yl)-2-(4-[1,3,4-oxadiazol-2-yl]phenyl)pyridine-3-sulfonamide, ZD4054

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
CHF 97.30
25 MG
CHF 392.00

CHF 97.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
CHF 97.30
25 MG
CHF 392.00

About This Item

Empirical Formula (Hill Notation):
C19H16N6O4S
CAS Number:
Molecular Weight:
424.43
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

CHF 97.30


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

O=S(C1=C(C2=CC=C(C3=NN=CO3)C=C2)N=CC=C1)(NC4=NC=C(C)N=C4OC)=O

InChI

1S/C19H16N6O4S/c1-12-10-21-17(19(23-12)28-2)25-30(26,27)15-4-3-9-20-16(15)13-5-7-14(8-6-13)18-24-22-11-29-18/h3-11H,1-2H3,(H,21,25)

InChI key

FJHHZXWJVIEFGJ-UHFFFAOYSA-N

Gene Information

human ... EDNRA(1909)

General description

Zibotentan is an orally bioavailable, non-peptide ETA (endothelin) receptor-specific antagonist, with no activity against ETB receptor.[1]

Biochem/physiol Actions

Zibotentan has been found to inhibit cell proliferation, invasion and metastasis in cancer models. In clinical trials on patients with castration-resistant prostate cancer and bone metastasis, who were pain-free or had mild pain, zibotentan treatment resulted in better overall survival as compared to placebo.
Zibotentan is a potent and selective endothelin ET(A) receptor antagonist with an IC50 of 21 nM for the ETA receptor subtype and no binding observed even at 10,000 nM for ETB.
Zibotentan is a potent and selective endothelin ET(A) receptor antagonist.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A phase I study of zibotentan (ZD4054) in patients with metastatic, castrate-resistant prostate cancer.
Schelman WR et al
Investigational New Drugs, 29(1), 118-125 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service