Skip to Content
Merck
All Photos(1)

Key Documents

SML1285

Sigma-Aldrich

Febuxostat

98.5-102.0% (dry basis), powder, xanthine oxidase inhibitor

Synonym(s):

2-(3-Cyano-4-isobutoxyphenyl)-4-methyl-1,3-thiazole-5-carboxylic acid, Adenuric, Atenuri, Uloric

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
CHF 247.00

CHF 247.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
CHF 247.00

About This Item

Empirical Formula (Hill Notation):
C16H16N2O3S
CAS Number:
Molecular Weight:
316.37
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

CHF 247.00


Check Cart for Availability

Request a Bulk Order

Product Name

Febuxostat, 98.5-102.0%

Quality Level

Assay

98.5-102.0%

form

powder

storage temp.

2-8°C

SMILES string

CC(C)COC1=C(C#N)C=C(C2=NC(C)=C(C(O)=O)S2)C=C1

InChI

1S/C16H16N2O3S/c1-9(2)8-21-13-5-4-11(6-12(13)7-17)15-18-10(3)14(22-15)16(19)20/h4-6,9H,8H2,1-3H3,(H,19,20)

InChI key

BQSJTQLCZDPROO-UHFFFAOYSA-N

General description

Febuxostat is a potent, non-purine compound, which inhibits the expression of cytokines/chemokines. It has also been reported to inhibit LPS-induced TNF-α, VCAM-1, MMP9 and MCP-1 expression.[1]

Biochem/physiol Actions

Febuxostat is a potent non-purine xanithine oxidase inhibitor.
Febuxostat is a potent non-purine xanithine oxidase inhibitor. Febuxostat is used in urate lowering therapies (ULTs) for the treatment of gout.

Caution

Freely soluble in N,N-dimethyl formamide, Soluble in dimethyl sulfoxide, sparingly soluble in ethanol or trichloromethane, slightly soluble in methanol, practically insoluble in water

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshiro Tanaka et al.
Free radical biology & medicine, 162, 298-308 (2021-01-21)
Accumulating evidence suggests that high serum uric acid (UA) is associated with left ventricular (LV) dysfunction. Although xanthine oxidase (XO) activation is a critical regulatory mechanism of the terminal step in ATP and purine degradation, the pathophysiological role of cardiac
Johji Nomura et al.
PloS one, 8(9), e75527-e75527 (2013-10-03)
Excess reactive oxygen species (ROS) formation can trigger various pathological conditions such as inflammation, in which xanthine oxidase (XO) is one major enzymatic source of ROS. Although XO has been reported to play essential roles in inflammatory conditions, the molecular

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service