Skip to Content
Merck
All Photos(1)

Key Documents

SML1062

Sigma-Aldrich

6-Aminophenanthridine

≥98% (HPLC)

Synonym(s):

6-Amino-phenanthridine, 6AP

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
CHF 84.90

CHF 84.90


Check Cart for Availability


Select a Size

Change View
5 MG
CHF 84.90

About This Item

Empirical Formula (Hill Notation):
C13H10N2
CAS Number:
Molecular Weight:
194.23
UNSPSC Code:
12352200
NACRES:
NA.77

CHF 84.90


Check Cart for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to brown

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

Nc1nc2c(c3c1cccc3)cccc2

InChI

1S/C13H10N2/c14-13-11-7-2-1-5-9(11)10-6-3-4-8-12(10)15-13/h1-8H,(H2,14,15)

InChI key

FVCXJXKLDUJOFA-UHFFFAOYSA-N

Biochem/physiol Actions

6-Aminophenanthridine selectively inhibits protein folding activity of the ribosome (PFAR).
6-Aminophenanthridine selectively inhibits protein folding activity of the ribosome (PFAR). Apparently 6-Aminophenanthridine interacts with the ribosome domain V rRNA and the protein substrate. 6-Aminophenanthridine is an antiprion compound that promotes mammalian prion clearance.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service