Skip to Content
Merck
All Photos(1)

Key Documents

SML0792

Sigma-Aldrich

JW480

≥98% (HPLC)

Synonym(s):

2-Isopropylphenyl (2-(naphthalen-2-yl)ethyl)carbamate, JW 480

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
CHF 94.00
25 MG
CHF 460.00

CHF 94.00


Estimated to ship on05 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
CHF 94.00
25 MG
CHF 460.00

About This Item

Empirical Formula (Hill Notation):
C22H23NO2
CAS Number:
Molecular Weight:
333.42
UNSPSC Code:
12352200
NACRES:
NA.77

CHF 94.00


Estimated to ship on05 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

N(CCc2cc3c(cc2)cccc3)C(=O)Oc1c(cccc1)C(C)C

InChI

1S/C22H23NO2/c1-16(2)20-9-5-6-10-21(20)25-22(24)23-14-13-17-11-12-18-7-3-4-8-19(18)15-17/h3-12,15-16H,13-14H2,1-2H3,(H,23,24)

InChI key

PCNJGBMWAZRVEA-UHFFFAOYSA-N

Biochem/physiol Actions

JW480 is a highly potent and selective inhibitor of KIAA1363.
JW480 is a highly potent and selective inhibitor of KIAA1363. JW480 reduce MAGEs and impair the migration, invasion, survival, and in vivo tumor growth of human prostate cancer cell lines.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuchuan Li et al.
Cancer cell international, 23(1), 276-276 (2023-11-18)
Despite therapeutic advances, the prognosis of pancreatic ductal adenocarcinoma (PDAC) remains extremely poor. Metabolic reprogramming is increasingly recognized as a key contributor to tumor progression and therapy resistance in PDAC. One of the main metabolic changes essential for tumor growth

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service