Skip to Content
Merck
All Photos(1)

Key Documents

SML0775

Sigma-Aldrich

PZ-II-029

≥98% (HPLC)

Synonym(s):

2,5-Dihydro-7-methoxy-2-(4-methoxyphenyl)-3H-Pyrazolo[4,3-c]quinolin-3-one, 7-Methoxy-2-(4-methoxyphenyl)-2H-pyrazolo[4,3-c]quinolin-3(5H)-one, PZII-029

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
CHF 354.00

CHF 354.00


Estimated to ship on25 April 2025


Request a Bulk Order

Select a Size

Change View
25 MG
CHF 354.00

About This Item

Empirical Formula (Hill Notation):
C18H15N3O3
CAS Number:
Molecular Weight:
321.33
UNSPSC Code:
12352200
NACRES:
NA.77

CHF 354.00


Estimated to ship on25 April 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

PZ-II-029 is an a6b3g2/3g2-specific GABAA positive modulator.
PZ-II-029 is the first compound to be identified as a specific postitive modulator of GABAA channels composed of α6β3γ2 subunits.

Features and Benefits

This compound is featured on the GABAA Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to GABAA receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service