Skip to Content
Merck
All Photos(1)

Key Documents

SML0286

Sigma-Aldrich

BMS 753

≥98% (HPLC)

Synonym(s):

4-[[(2,3-Dihydro-1,1,3,3-tetramethyl-2-oxo-1H-inden-5-yl)carbonyl]amino]-benzoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
CHF 110.00

CHF 110.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
5 MG
CHF 110.00

About This Item

Empirical Formula (Hill Notation):
C21H21NO4
CAS Number:
Molecular Weight:
351.40
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

CHF 110.00


Estimated to ship on02 June 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: ≥5 mg/mL

storage temp.

2-8°C

SMILES string

CC1(C)C(=O)C(C)(C)c2cc(ccc12)C(=O)Nc3ccc(cc3)C(O)=O

InChI

1S/C21H21NO4/c1-20(2)15-10-7-13(11-16(15)21(3,4)19(20)26)17(23)22-14-8-5-12(6-9-14)18(24)25/h5-11H,1-4H3,(H,22,23)(H,24,25)

InChI key

KFBPBWUZXBYJDG-UHFFFAOYSA-N

Biochem/physiol Actions

BMS 753 is a very potent, specific agonist for RARa (Ki = 2 nM).
BMS 753 is a very potent, specific agonist for RARa (Ki = 2 nM). BMS 753 does not display significant effects on RARg in reporter based assays, or in binding assays measuring displacement of labeled retinoic acid.

Features and Benefits

This compound is featured on the Nuclear Receptors (Non-Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to non-steroid nuclear receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service