SML0121
BTZO-1
≥98% (HPLC)
Synonym(s):
2-Pyridin-2-yl-4H-1,3-benzothiazin-4-one
About This Item
Recommended Products
Assay
≥98% (HPLC)
form
powder
color
white to tan
solubility
DMSO: ≥7 mg/mL
originator
Takeda
storage temp.
2-8°C
SMILES string
O=C1N=C(Sc2ccccc12)c3ccccn3
InChI
1S/C13H8N2OS/c16-12-9-5-1-2-7-11(9)17-13(15-12)10-6-3-4-8-14-10/h1-8H
InChI key
GBAKVEWPYUIGHN-UHFFFAOYSA-N
Application
Biochem/physiol Actions
Features and Benefits
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service