Skip to Content
Merck
All Photos(1)

Key Documents

SML0116

Sigma-Aldrich

GW788388 Hydrate

≥98% (HPLC)

Synonym(s):

4-{4-[3-(Pyridin-2-yl)-1H-pyrazol-4-yl]-pyridin-2-yl}-N-(tetrahydro-2H-pyran-4-yl)benzamide hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
CHF 165.00

CHF 165.00


Check Cart for Availability


Select a Size

Change View
5 MG
CHF 165.00

About This Item

Empirical Formula (Hill Notation):
C25H23N5O2 · xH2O
Molecular Weight:
425.48 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

CHF 165.00


Check Cart for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥8 mg/mL

originator

GlaxoSmithKline

storage temp.

2-8°C

SMILES string

O.O=C(NC1CCOCC1)c2ccc(cc2)-c3cc(ccn3)-c4c[nH]nc4-c5ccccn5

InChI

1S/C25H23N5O2.H2O/c31-25(29-20-9-13-32-14-10-20)18-6-4-17(5-7-18)23-15-19(8-12-27-23)21-16-28-30-24(21)22-3-1-2-11-26-22;/h1-8,11-12,15-16,20H,9-10,13-14H2,(H,28,30)(H,29,31);1H2

InChI key

VZRJMBZMFKADGR-UHFFFAOYSA-N

Biochem/physiol Actions

GW788388 is a potent selective inhibitor of TGF-beta type I receptor [activin receptor-like kinase 5 (ALK5)]. GW788388 is orally available with a half-life of approximately 4 hours.
TGFβ1 (ALK5) inhibitor; Potent, selective inhibitor of TGFβ signalling in vitro and renal fibrosis in vivo

Features and Benefits

This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Min Cheol Pyo et al.
Archives of toxicology, 94(9), 3329-3342 (2020-07-04)
Ochratoxin A (OTA) is a toxin produced by fungi such as Aspergillus spp. and Penicillium spp. The key target organ of OTA toxicity is the kidney, and it is known that epithelial-to-mesenchymal transition (EMT) leading to fibrosis is enhanced after
Hsiu-Hui Yang et al.
Frontiers in physiology, 13, 816959-816959 (2022-06-11)
Water permeability of the kidney collecting ducts is regulated in part by the amount of the molecular water channel protein aquaporin-2 (AQP2), whose expression, in turn, is regulated by the pituitary peptide hormone vasopressin. We previously showed that stable glucocorticoid
Yunmee Lho et al.
International journal of molecular sciences, 22(9) (2021-05-06)
We investigated the effectiveness of the transforming growth factor beta-1 (TGF-β) receptor inhibitor GW788388 on the epithelial to mesenchymal transition (EMT) using human peritoneal mesothelial cells (HPMCs) and examined the effectiveness of GW788388 on the peritoneal membrane using a peritoneal
Paul Jennings et al.
Cell biology and toxicology (2023-01-01)
Transcriptomic analysis is a powerful method in the utilization of New Approach Methods (NAMs) for identifying mechanisms of toxicity and application to hazard characterization. With this regard, mapping toxicological events to time of exposure would be helpful to characterize early

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service