Skip to Content
Merck
All Photos(1)

Key Documents

SML0081

Sigma-Aldrich

Zacopride hydrochloride hydrate

≥98% (HPLC)

Synonym(s):

4-Amino-N-1-azabicyclo[2.2.2]oct-3-yl-5-chloro-2-methoxybenzamide hydrochloride hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
CHF 147.00
50 MG
CHF 592.00

CHF 147.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
CHF 147.00
50 MG
CHF 592.00

About This Item

Empirical Formula (Hill Notation):
C15H20ClN3O2 · HCl · xH2O
CAS Number:
Molecular Weight:
346.25 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

CHF 147.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to tan

solubility

H2O: ≥10 mg/mL

originator

Wyeth

storage temp.

2-8°C

SMILES string

O.Cl.COc1cc(N)c(Cl)cc1C(=O)NC2CN3CC[C@H]2CC3

InChI

1S/C15H20ClN3O2.ClH.H2O/c1-21-14-7-12(17)11(16)6-10(14)15(20)18-13-8-19-4-2-9(13)3-5-19;;/h6-7,9,13H,2-5,8,17H2,1H3,(H,18,20);1H;1H2

InChI key

NPXXKMBXEODDAF-UHFFFAOYSA-N

Biochem/physiol Actions

Serotonin 5-HT3 and 5-HT4 receptor antagonist
Zacopride is a sub-nanomolar antagonist of the serotonin 5-HT3 receptor that also displays agonist-like activity against 5-HT4 receptors. It is a potent anxiolytic in animal models, and also has strong antiemetic effects, and also increases aldosterone levels via activation of 5-HT4 receptor in the adrenal glands.

Features and Benefits

This compound was developed by Wyeth. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service