Skip to Content
Merck
All Photos(1)

Key Documents

SMB01079

Sigma-Aldrich

Ambrosic acid

≥90% (LC/MS-ELSD)

Synonym(s):

2-(5,10-dimethyl-4-oxo-11-oxatricyclo[6.2.1.01,5]undecan-7-yl)prop-2-enoic acid, Peruvic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
CHF 294.00

CHF 294.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
CHF 294.00

About This Item

Empirical Formula (Hill Notation):
C15H20O4
CAS Number:
Molecular Weight:
264.32
UNSPSC Code:
12352205
NACRES:
NA.25

CHF 294.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

264.32

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O1C32C(CC(C1CC3C)C(=C)C(=O)O)(C(=O)CC2)C

InChI key

ZONXEWIGPLHXNT-UHFFFAOYSA-N

General description

Ambrosic acid, also known as peruvic acid, is a member of oxanes, a natural product commonly found in Ambrosia tenuifolia and Ambrosia artemisiifolia. It is a sesquiterpenoid[1] with antioxidant and neuroprotective activities[2].

Application

It is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical and biochemical research.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sesquiterpenoids from the Inflorescence of Ambrosia artemisiifolia
Zheng Z, et al.
Molecules (Basel), 27, 5915-5915 (2022)
Eudesmane Glycosides from Ambrosia artemisiifolia (Common Ragweed) as Potential Neuroprotective Agents
An JP, et al.
Journal of Natural Products, 82, 1128-1138 (2019)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service