Skip to Content
Merck
All Photos(1)

Key Documents

SMB01067

Sigma-Aldrich

Angeloylgomisin H

≥90% (LC/MS-ELSD)

Synonym(s):

(+)-Angeloylgomisin H, tigloylgomisin H

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
CHF 326.00

CHF 326.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
CHF 326.00

About This Item

Empirical Formula (Hill Notation):
C28H36O8
CAS Number:
Molecular Weight:
500.58
UNSPSC Code:
12352205
NACRES:
NA.25

CHF 326.00


Check Cart for Availability

Request a Bulk Order

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

500.58

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

OC1(C(Cc2c(c(c(c(c2)OC)OC)OC(=O)C(=CC)C)c3c(cc(c(c3OC)OC)OC)C1)C)C

InChI

1S/C28H36O8/c1-10-15(2)27(29)36-26-21-17(12-19(31-5)24(26)34-8)11-16(3)28(4,30)14-18-13-20(32-6)23(33-7)25(35-9)22(18)21/h10,12-13,16,30H,11,14H2,1-9H3

InChI key

ZSAUXCVJDYCLRS-UHFFFAOYSA-N

General description

Angeloylgomisin H, a lignan, is a natural product commonly found in the fruits of Schisandra chinensis and the roots of Angelica Dahurica. This compound exhibits potent biological activity, including anti-inflammatory, antioxidant, antidiabetic, antiproliferative, and anticancer properties.

Application

It is a natural product derived from plant source that finds application in screening, metabolomics, phytochemical and pharmaceutical research.

Biochem/physiol Actions

  • Inhibits the activity of enzymes involved in carcinogenic metabolism, such as ornithine decarboxylase and acetylcholinesterase.
  • Facilitates the detoxification of specific carcinogens, including benzo(a)pyrene, by converting them into non-toxic metabolites.
  • Exhibits anti-diabetic effects, potentially linked to its capacity to stimulate insulin release.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service