Skip to Content
Merck
All Photos(1)

Documents

SIAL0790

Sigma-Aldrich

Sigma® centrifuge tubes

capacity 15 mL, sterile, pack of 10 × 50 ea racked

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122804
NACRES:
NB.22

material

polypropylene

description

marking spot

sterility

sterile

feature

RCF 12500 × g
disposable
graduated
graduations 1.5 ‑ 14.5 mL

packaging

pack of 10 × 50 ea racked

O.D. × L

17.5 mm × 118.5 mm

cap diam.

22.6 mm

capacity

15 mL

color

natural

Looking for similar products? Visit Product Comparison Guide

General description

Sigma disposable 15 mL centrifuge tubes are available as clear polyethylene terephthalate (PET) tubes with a traditional plug seal cap or polypropylene (PP) tubes with either plug seal caps or the new easy-on/easy-off top offering advanced ergonomics with its wider knurls and roll-over edge design. The Sigma centrifuge tubes are available as polypropylene tubes either racked or bulk packed.

  • Sterile, certified nonpyrogenic and DNase-/RNase-free
  • Black printed graduations and a large white marking spot
  • Maximum RCF for PET tubes is 3,600xg

Legal Information

Sigma is a registered trademark of Sigma-Aldrich Co. LLC

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Universal Transfection Reagent enables efficient nucleic acid delivery into various cells, compatible with different cell culture conditions.

3D Spheroid BME Cell Invasion Assay utilizes specialized plates and ECM for cell aggregation and spheroid formation in vitro.

3-D Culture Matrix™ Collagen I facilitates cellular growth and differentiation in vitro, offering versatile applications in biomedical research.

Calcium phosphate transfection is a common method for the introduction of DNA into eukaryotic cells. This protocol can be optimized for use with a wide variety of cell types.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service