Skip to Content
Merck
All Photos(1)

Key Documents

SCP0247

Sigma-Aldrich

Factor VIIa substrate

≥95% (HPLC), lyophilized

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
CHF 188.00

CHF 188.00


Check Cart for Availability


Select a Size

Change View
5 MG
CHF 188.00

About This Item

Empirical Formula (Hill Notation):
C30H42N7O7S1
Molecular Weight:
644.76
UNSPSC Code:
12352204
NACRES:
NA.32

CHF 188.00


Check Cart for Availability

Product Name

Factor VIIa substrate,

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥72%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

MeSO2-Cha-Abu-Arg-AFC

Application

Factor VIIa is a serine protease involved in the blood coagulation cascade. The activity of Factor VIIa can be assayed with chromogenic substrate MeSO2-Cha-Abu-Arg-pNA or the flourogenic substrate MeSO2-Cha-Abu-Arg-AFC.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giuseppe Lippi et al.
Seminars in thrombosis and hemostasis, 35(1), 9-22 (2009-03-25)
Hemostasis is traditionally defined as the physiologic process whereby bleeding is antagonized and possibly stopped to minimize blood loss. The first medical description of the clinical and inherited features of hemostasis can be dated back more than 1000 years, when
Flora Peyvandi et al.
Thrombosis and haemostasis, 88(5), 750-755 (2002-11-13)
Factor VII (FVII) requires the cleavage of an internal peptide bond and the association with tissue factor (TF) to attain its fully active FVIIa conformation. This event alone leaves FVIIa in a zymogen-like state of relatively low specific activity. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service