Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Select a Size
CHF 281.00
Select a Size
About This Item
CHF 281.00
Recommended Products
Product Name
MMP 8 Substrate, ≥95% (HPLC)
Assay
≥95% (HPLC)
form
lyophilized
composition
Peptide Content, ≥80%
storage condition
protect from light
storage temp.
−20°C
Amino Acid Sequence
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
What is the Department of Transportation shipping information for this product?
1 answer-
Helpful?
-
-
How is product SCP0188, MMP 8 Substrate, used for detecting MMP-8 activity?
1 answer-
The use of DNP-Pro-Leu-Ala-Tyr-Trp-Ala-Arg as a fluorogenic substrate in measuring the activity of human neutrophil collagenase (MMP-8) is described in Netzel-Arnett, S., et al., Continuously recordingfluorescent assays optimized for five human matrix metalloproteinases. Anal. Biochem., 195, 86-92 (1991).
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service