All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C38H59N9O13
Molecular Weight:
849.93
UNSPSC Code:
12352200
NACRES:
NA.32
Assay
≥95% (HPLC)
form
lyophilized
composition
Peptide Content, ≥68%
storage condition
protect from light
storage temp.
−20°C
Amino Acid Sequence
Tyr-Gly-Ala-Val-Val-Asn-Asp-Leu
Application
The octapeptide YGAVVNDL is closely related to the nonapeptide YAGAVVNDL used to inhibit herpes virus ribonucleotide reductase.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
J Furlong et al.
Virology, 182(2), 846-851 (1991-06-01)
The open reading frame of the large subunit (R1) of herpes simplex virus type 1 (HSV-1) ribonucleotide reductase has been positioned downstream of the phage T7 gene 10 promoter in the expression vector, pET. Transformation of this recombinant plasmid into
A Marcello et al.
Journal of chemotherapy (Florence, Italy), 7(5), 403-405 (1995-10-01)
Peptides capable of selectively disrupting protein-protein interactions that are required for viral replication represent potential agents for antiviral therapy. The first example of viral product that could be inhibited by the peptide YAGAVVNDL, targeted to the functional interaction between subunits
H Paradis et al.
The Journal of biological chemistry, 263(31), 16045-16050 (1988-11-05)
Herpes simplex virus (HSV) ribonucleotide reductase activity is specifically inhibited by a synthetic peptide, Tyr-Ala-Gly-Ala-Val-Val-Asn-Asp-Leu (HSV H2-(7-15], corresponding to the carboxyl terminus of its subunit 2 (H2). In order to elucidate the mechanism of action of the nonapeptide a photoreactive
A Marcello et al.
Proceedings of the National Academy of Sciences of the United States of America, 91(19), 8994-8998 (1994-09-13)
Mimetic peptides capable of selectively disrupting protein-protein interactions represent potential therapeutic agents for inhibition of viral and cellular enzymes. This approach was first suggested by the observation that the peptide YAGAVVNDL, corresponding to the carboxyl-terminal 9 amino acids of the
E Telford et al.
The Journal of general virology, 71 ( Pt 6), 1373-1378 (1990-06-01)
The synthetic nonapeptide YAGAVVNDL [identical to the nine carboxy-terminal amino acids of the small subunit of herpes simplex virus (HSV)-encoded ribonucleotide reductase (RR)] was found to inhibit the RR activity induced by equine herpesvirus type 1 subtype 1 (EHV-1). Parallel
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service