Skip to Content
Merck
All Photos(1)

Documents

SAE0153

Sigma-Aldrich

Leupeptin Ready Made Solution

Non-freezing solution, 5 mg/ml

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.54

biological source

microbial

Quality Level

Assay

≥90% (HPLC)

form

ready-to-use solution

concentration

5 mg/mL

storage temp.

−20°C

General description

Leupeptin is a tripeptide protease inhibitor that is naturally produced in Actinomyces / Streptomyces. Known also as N-acetyl-L-leucyl-L-leucyl-L-arginal, leupeptin is a reversible competitive inhibitor of cysteine, serine, and thiol proteases.

Features and Benefits

  • Inhibitor of serine and cysteine proteases
  • Inhibits plasmin, trypsin, papain, calpain, and cathepsin B
  • Does not inhibit pepsin, cathepsin A, cathepsin D, thrombin, or α-chymotrypsin
  • Effective concentration: 10 μM - 100 μM
  • Convenient 5 mM non-freezing solution, ready to dilute to the desired working concentration

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service