Skip to Content
Merck
All Photos(2)

Key Documents

SAB5700317

Sigma-Aldrich

Anti-Phospho-MDM2-S166 antibody produced in rabbit

Synonym(s):

ACTFS, HDMX, MDM2, hdm2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CHF 365.00

CHF 365.00


Check Cart for Availability


Select a Size

Change View
100 μL
CHF 365.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 365.00


Check Cart for Availability

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

11-14
24-55

species reactivity

rat, mouse, human

concentration

0.63 mg/ml

technique(s)

immunofluorescence: 1:50-1:100
western blot: 1:500-1:1000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer166)

Gene Information

human ... MDM2(4193)

General description

This gene encodes a nuclear-localized E3 ubiquitin ligase. The encoded protein can promote tumor formation by targeting tumor suppressor proteins, such as p53, for proteasomal degradation. This gene is itself transcriptionally-regulated by p53. Overexpression or amplification of this locus is detected in a variety of different cancers. There is a pseudogene for this gene on chromosome 2. Alternative splicing results in a multitude of transcript variants, many of which may be expressed only in tumor cells.

Immunogen

A synthetic phosphorylated peptide around S166 of human MDM2 (NP_002383.2).

Physical form

PBS with 0.02% sodium azide,50% glycerol,pH7.3.

Storage and Stability

Store at -20℃. Avoid freeze / thaw cycles.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service