Skip to Content
Merck
All Photos(2)

Key Documents

SAB5500153

Sigma-Aldrich

Anti-N-RAS antibody, Rabbit monoclonal

recombinant, expressed in proprietary host, clone SP174, affinity isolated antibody

Synonym(s):

Anti-ALPS4, Anti-CMNS, Anti-KRAS, Anti-N-ras, Anti-NCMS, Anti-NRAS1, Anti-NS6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in proprietary host

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

SP174, monoclonal

species reactivity

human (tested)

technique(s)

immunoblotting: 1:400
immunohistochemistry: 1:100

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... NRAS(4893)

General description

GTPase N-Ras or N-Ras is a guanine-nucleotide binding protein. Receptor tyrosine kinases and G-protein-coupled receptors activate Ras, which then stimulates the MAPK signal pathway. Point mutations (Q61R) in Ras prevent the GAP-mediated inhibition of the MAPK pathway in certain cancers such as lung carcinoma and melanoma. Abnormal MAPK signaling may lead to uncontrolled cell proliferation, resistance to both apoptosis and cancer therapy.

Immunogen

Synthetic peptide of human N-Ras protein containing N-Ras (Q61R) point mutation.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

0.1 ml rabbit monoclonal antibody purified by protein A/G in PBS/1% BSA buffer pH 7.6 with less than 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service