Skip to Content
Merck
All Photos(1)

Key Documents

SAB4701036

Sigma-Aldrich

Monoclonal Anti-AGR2/AGR3 antibody produced in mouse

clone AGR3.4

Synonym(s):

AG2, AG3, HAG2, HAG3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 380.00

CHF 380.00


Estimated to ship on27 April 2025



Select a Size

Change View
100 μG
CHF 380.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 380.00


Estimated to ship on27 April 2025


biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

AGR3.4, monoclonal

form

buffered aqueous solution

concentration

1 mg/mL

technique(s)

western blot: 1 μg/mL

accession no.

O95994

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... AGR2(10551)

General description

The antibody AGR3.4 recognizes the PLMII epitope of AGR3 (AG3) and AGR2 (AG2) proteins (19-20 kDa); secreted cytoplasmic proteins which can serve as markers of carcinogenesis.

Immunogen

Purified human AGR3 protein

Physical form

Solution in phosphate buffered saline with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service