Skip to Content
Merck
All Photos(1)

Key Documents

SAB4700763

Sigma-Aldrich

Monoclonal Anti-PAG / Cbp antibody produced in mouse

clone PAG-C1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-Cbp

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 254.00

CHF 254.00


Estimated to ship on26 May 2025



Select a Size

Change View
100 μG
CHF 254.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 254.00


Estimated to ship on26 May 2025


biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

PAG-C1, monoclonal

form

buffered aqueous solution

species reactivity

mouse, rat, human, bovine

concentration

1 mg/mL

technique(s)

immunoblotting: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG2b

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... PAG1(55824)

General description

The antibody PAG-C1 recognizes an epitope located in the C-terminal domain of Csk-binding protein (Cbp / PAG), a 46 kDa ubiquitously expressed transmembrane adaptor protein present in membrane rafts (glycosphingolipid-enriched microdomains), which however migrates on SDS PAGE gels anomalously as an 80 kDa molecule.

Immunogen

C-terminal peptide (last 15 amino acids) of human Csk binding protein coupled to KLH.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service