Skip to Content
Merck
All Photos(1)

Documents

SAB4700352

Sigma-Aldrich

Monoclonal Anti-IgE antibody produced in mouse

clone 4H10, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-IGHE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4H10, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... IGHE(3497)

Related Categories

General description

Immunoglobulin E (IgE) is encoded by the gene mapped to human chromosome 14q32.33. IgE belongs to the immunoglobulin family and consists of epsilon(ε) heavy chain in the constant (C) region. IgE has a monomeric structure with an extra domain in the constant region.
The antibody 4H10 reacts with human IgE; it recognizes an epitope different from the ones recognized by BE5 and 4G7 antibodies to IgE.

Immunogen

Purified human IgE

Application

The reagent is designed for Flow Cytometry analysis. Suggested working dilution for Flow Cytometry is 4 μg/mL of sample. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Biochem/physiol Actions

Immunoglobulin E (IgE) binds very tightly to Fc receptors on basophils and mast cells.Thus, it plays a major role in stimulating allergic reactions. Elevated expression of the gene has been associated with the development of hyper IgE Syndrome (HIES), hay fever, atopic skin diseases such as eczema, anaphylactic shock and IgE-myeloma. In addition, overexpression of IgE is also observed in allergy/asthma.Therefore, increased levels of serum IgE can be considered as a diagnosis for asthma. IgE expression is down-regulated in congenital agammaglobulinemia and hypogammaglobulinemia because of faulty metabolism or synthesis of immunoglobulins.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in Tris buffered saline, pH 8.0, with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service