Skip to Content
Merck
All Photos(1)

Key Documents

SAB4503368

Sigma-Aldrich

Anti-Collagen XXV α1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

AMY, Alzheimer disease amyloid-associated protein, CLAC-P, Collagen α-1(XXV) chain, Collagen-like Alzheimer amyloid plaque component

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 389.00

CHF 389.00


Estimated to ship on29 May 2025



Select a Size

Change View
100 μG
CHF 389.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 389.00


Estimated to ship on29 May 2025


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 64 kDa

species reactivity

human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... COL25A1(84570)

General description

Anti-Collagen XXV α1 Antibody detects endogenous levels of total Collagen XXV α1 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human Collagen XXV alpha1.

Immunogen Range: 101-150

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service