Skip to Content
Merck
All Photos(3)

Key Documents

SAB4300350

Sigma-Aldrich

Anti-ERBB2 (Ab-1221/1222) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CD340 antibody produced in rabbit, Anti-HER-2 antibody produced in rabbit, Anti-HER-2/neu antibody produced in rabbit, Anti-HER2 antibody produced in rabbit, Anti-v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro / glioblastoma derived oncogene homolog (avian) antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 399.00

CHF 399.00


Estimated to ship on23 April 2025



Select a Size

Change View
100 μG
CHF 399.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 399.00


Estimated to ship on23 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~185 kDa

species reactivity

human

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
indirect immunofluorescence: 1:100-1:200
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(N-L-Y-Y-W)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ERBB2(2064)

Related Categories

General description

ERBB2 (erb-b2 receptor tyrosine kinase 2) is a tyrosine kinase receptor that belongs to the ErbB family. It is also known as HER2/Neu. It is located on chromosome 17q12.

Immunogen

Peptide sequence around aa. 1219-1223/1220-1224 (N-L-Y-Y -W), according to the protein ERBB2.

Biochem/physiol Actions

ERBB2 (erb-b2 receptor tyrosine kinase 2) is considered as a therapeutic target in breast cancer. It controls the concentration of intracellular calcium in SKBR3 (breast cancer cell line) cells.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Essential component of a neuregulin-receptor complex, although neuregulins do not interact with it alone. GP30 is a potential ligand for this receptor. Not activated by EGF, TGF-alpha and amphiregulin.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HER2 signaling regulates HER2 localization and membrane retention
Jeong J, et al.
PLoS ONE (2017)
Detection and mapping of amplified DNA sequences in breast cancer by comparative genomic hybridization
Kallioniemi A, et al.
Proceedings of the National Academy of Sciences of the USA (1994)
The HER2 phenotype of circulating tumor cells in HER2-positive early breast cancer: A translational research project of a prospective randomized phase III trial
Jaeger BAS, et al.
PLoS ONE (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service