Skip to Content
Merck
All Photos(2)

Key Documents

SAB4300279

Sigma-Aldrich

Anti-phospho-CDK6 (pTyr13) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-MGC59692 antibody produced in rabbit, Anti-PLSTIRE antibody produced in rabbit, Anti-STQTL11 antibody produced in rabbit, Anti-cyclin-dependent kinase 6 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 347.00

CHF 347.00


Check Cart for Availability


Select a Size

Change View
100 μG
CHF 347.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 347.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~36 kDa

species reactivity

human, mouse, rat

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
indirect immunofluorescence: 1:100-1:200

isotype

IgG

immunogen sequence

(Q-Q-YP-E-C)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr13)

Gene Information

human ... CDK6(1021)

Immunogen

Peptide sequence around phosphorylation site of tyrosine 13 (Q-Q-Y(p)-E-C), according to the protein CDK6.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Probably involved in the control of the cell cycle. Interacts with D-type G1 cyclins.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service