Skip to Content
Merck
All Photos(1)

Documents

SAB4300092

Sigma-Aldrich

Anti-phospho-RPS6KA1 (pSer352) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-HU-1 antibody produced in rabbit, Anti-MAPKAPK1A antibody produced in rabbit, Anti-RSK antibody produced in rabbit, Anti-RSK1 antibody produced in rabbit, Anti-ribosomal protein S6 kinase, 90kDa, polypeptide 1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~90 kDa

species reactivity

human, rat, mouse

concentration

1 mg/mL

technique(s)

western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(R-D-SP-P-G)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... RPS6KA1(6195)

Immunogen

Peptide sequence around phosphorylation site of serine 352 (R-D-S(p)-P-G), according to the protein RPS6KA1.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Serine/threonine kinase that may play a role in mediating the growth-factor and stress induced activation of the transcription factor CREB.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isabelle Re Nett et al.
EMBO reports, 19(8) (2018-06-14)
Mitogen-activated protein kinase (MAPK)/extracellular signal-regulated kinase (ERK) signalling is implicated in initiation of embryonic stem (ES) cell differentiation. The pathway is subject to complex feedback regulation. Here, we examined the ERK-responsive phosphoproteome in ES cells and identified the negative regulator
B Nadratowska-Wesolowska et al.
Oncogene, 33(40), 4823-4836 (2013-10-22)
FGFR1 (fibroblast growth factor receptor 1) regulates many key cellular responses including proliferation, migration and differentiation through activation of signaling pathways. Irregularities in FGFR1 signaling have been implicated in several pathological conditions, including human cancer. In order to discover novel

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service