Skip to Content
Merck
All Photos(1)

Documents

SAB4200824

Sigma-Aldrich

Anti-TFE3 antibody, Mouse monoclonal

clone TFE3-37, purified from hybridoma cell culture

Synonym(s):

Class E basic helix-loop-helix protein 33, Transcription factor E3, bHLHe33

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

antibody form

purified from hybridoma cell culture

clone

TFE3-37, monoclonal

form

buffered aqueous solution

mol wt

60 kDa

species reactivity

human, mouse, bovine, rat, canine

packaging

antibody small pack of 25 μL

concentration

~1 mg/mL

technique(s)

immunoblotting: 1.5-3 μg/mL using human A549 cells extract

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TFE3(7030)

Related Categories

General description

Transcription factor E3 (TFE3), also known as Class E basic helix-loop-helix protein 33 (bHLHe33), is a transcriptional activator that mediates the enhancer-promoter interactions. TFE3 belongs to the MiT family of helix-loop-helix leucine zipper transcription factors, it is ubiquitously expressed and can directly associate with DNA as either homodimer or heterodimer which are formed with two other MiT family members, TFEB or TFEC.
TFE3 serve an important role in cell growth, cell proliferation, cellular adaptation to starvation and cellular response to ER stress. Under nutrient-rich conditions, TFE3 is located in the cytoplasm, and under starvation conditions or treatment with ER stressors, TFE3 rapidly translocates to the nuclear. In the nuclear localization, TFE3 mediates cellular adaptation to stress by simultaneously promoting lysosomal biogenesis, autophagy induction, as well as expression of critical mitochondrial and metabolic regulators. It been shown that TFE3 also participate in the transcriptional regulation of the innate immune response.
Pathogen infections promote TFE3 nuclear translocation, thus inducing In vivo expression of several cytokines and chemokines.

Application

Anti-TFE3 antibody specifically recognizes TFE3 from human, mouse, rat, bovine and canine origin. The antibody may be used in various immunochemical techniques including Immunoblotting (~60 kDa). Detection of the TFE3 band by Immunoblotting is specifically inhibited by the immunizing peptide.

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide as a preservative.

Other Notes

Unless otherwise stated in our catalog our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Disclaimer

This product is for R&D use only, not for drug, household, or other uses.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

José A Martina et al.
Science signaling, 7(309), ra9-ra9 (2014-01-23)
The discovery of a gene network regulating lysosomal biogenesis and its transcriptional regulator transcription factor EB (TFEB) revealed that cells monitor lysosomal function and respond to degradation requirements and environmental cues. We report the identification of transcription factor E3 (TFE3)
Nunzia Pastore et al.
Autophagy, 12(8), 1240-1258 (2016-05-14)
The activation of transcription factors is critical to ensure an effective defense against pathogens. In this study we identify a critical and complementary role of the transcription factors TFEB and TFE3 in innate immune response. By using a combination of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service