Skip to Content
Merck
All Photos(4)

Key Documents

SAB4200611

Sigma-Aldrich

Anti-DKC1 antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-CBF5, Anti-CBF5 homolog, Anti-DKC, Anti-H/ACA ribonucleoprotein complex subunit 4, Anti-NAP57, Anti-NOLA4, Anti-XAP101, Anti-cbf5p homolog, Anti-dyskeratosis congenita 1, Anti-dyskerin, Anti-nopp140-associated protein of 57 kDa, Anti-nucleolar protein family A member 4, Anti-snoRNP protein DKC1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~57 kDa

species reactivity

human, mouse

concentration

~1.0 mg/mL

technique(s)

immunoblotting: 0.5-1.0 μg/mL using whole extracts of mouse Hepa1-6 cells or nuclear extract of HeLa cells.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DKC1(1736)

General description

Dyskerin is a nucleolar protein encoded by the DKC1 gene. This gene is a member of the H/ACA snoRNPs (small nucleolar ribonucleoproteins) gene family. The DKC1 gene lies in a tail-to-tail orientation with the palmitoylated erythrocyte membrane protein gene and is transcribed in a telomere to centromere direction.

Immunogen

peptide corresponding to the N-terminal region of human DKC1, conjugated to KLH. The corresponding sequence is identical in monkey, bovine, pig and dog and differs by a single amino acid in mouse and rat.

Application

Anti-DKC1 antibody has been used in immunoblotting.

Biochem/physiol Actions

Dyskerin and the three NOLA proteins localize to the dense fibrillar components of nucleoli and to coiled (Cajal) bodies in the nucleus. Mutations in the DKC1 gene cause X-linked dyskeratosis congenita, aplastic anemia and progressive bone marrow failure in most cases. It also leads to a disease resulting in reticulate skin pigmentation, mucosal leukoplakia, nail dystrophy. It also causes Hoyeraal-Hreidarsson syndrome, which is a more severe form of dyskeratosis congenita. The overexpression of dyskerin has been observed in many cancers including neuroblastoma, colorectal cancer, lymphoma, breast cancer, melanoma, prostate cancer, ovarian carcinoma and hepatocellular carcinoma.

Physical form

a solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dyskerin overexpression in human hepatocellular carcinoma is associated with advanced clinical stage and poor patient prognosis
Liu B, et al.
PLoS ONE, 7(8), e43147-e43147 (2012)
Telomerase reverse-transcriptase homozygous mutations in autosomal recessive dyskeratosis congenita and Hoyeraal-Hreidarsson syndrome
Marrone A, et al.
Blood, 110(13), 4198-4205 (2007)
An enhanced H/ACA RNP assembly mechanism for human telomerase RNA
Egan E D and Collins K
Molecular and Cellular Biology, 32(13), 2428-2439 (2012)
Mouse dyskerin mutations affect accumulation of telomerase RNA and small nucleolar RNA, telomerase activity, and ribosomal RNA processing
Mochizuki Y, et al.
Proceedings of the National Academy of Sciences of the USA, 101(29), 10756-10761 (2004)
Brendan J Quinn et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(47), 19842-19847 (2009-11-10)
As mediators of innate immunity, neutrophils respond to chemoattractants by adopting a highly polarized morphology. Efficient chemotaxis requires the formation of one prominent pseudopod at the cell front characterized by actin polymerization, while local inhibition suppresses the formation of rear

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service