Skip to Content
Merck
All Photos(3)

Key Documents

SAB4200458

Sigma-Aldrich

Anti-ApoD (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-Apolipoprotein D precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~29 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

immunohistochemistry: 20 μg/mL using formalin-fixed, paraffin-embedded human breast carcinoma
indirect immunofluorescence: 0.1-0.2 μg/mL using HEK-293T cells over-expressing human ApoD
western blot: 0.1-0.2 μg/mL using HEK-293T cells over-expressing human ApoD

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... APOD(347)

General description

Apoliporotein D (ApoD), a 29-kDa apolipoprotein bound to high density lipoproteins (HDL) in human plasma, is a member of the lipocalin superfamily of proteins. ApoD is widely expressed at high levels in a variety of tissues such as spleen, testes and brain (central nervous system). In the CNS, ApoD is mainly expressed by astrocytes and oligodendrocytes from late embryogenesis to aged nervous system.

Immunogen

synthetic peptide corresponding to a sequence at the C-terminus of human apolipoprotein-D, conjugated to KLH. The corresponding sequence has 71% sequence identity in rat and mouse ApoD.

Application

Anti-ApoD (C-terminal) antibody produced in rabbit has been used in several immunochemical techniques including immunoblotting, immunofluorescence and immunohistochemistry.

Biochem/physiol Actions

Apoliporotein D (ApoD) acts as transporters of small hydrophobic ligands.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Apolipoprotein D
Rassart E, et al.
Biochimica et Biophysica Acta, 1482, 185-198 (2000)
Apolipoprotein D expression in human brain reactive astrocytes
del Valle E, et al.
The Journal of Histochemistry and Cytochemistry, 51(10), 1285-1290 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service