Skip to Content
Merck
All Photos(1)

Key Documents

SAB4100248

Sigma-Aldrich

Monoclonal Anti-AQP1 antibody produced in mouse

clone AQP1-B217, culture supernatant

Synonym(s):

AntiAQP-CHIP, AntiAquaporin 1 (Colton blood group), AntiAquaporin 1 (channel-forming integral protein, 28kDa, CO blood group), AntiCHIP28, AntiCO, AntiChannel-like integral membrane protein, 28-kDa, AntiColton blood group, AntiFIX

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
CHF 347.00

CHF 347.00


Estimated to ship on01 June 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1390

Request a Bulk Order

Select a Size

Change View
200 μL
CHF 347.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 347.00


Estimated to ship on01 June 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1390

Request a Bulk Order

biological source

mouse

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

AQP1-B217, monoclonal

form

aqueous solution

mol wt

antigen predicted mol wt 28.3 kDa

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AQP1(358)

Immunogen

AQP1 purified recombinant protein

Physical form

Solution in Dulbecco′s Modified Eagle′s Medium with 10% fetal calf serum and 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service