Skip to Content
Merck
All Photos(1)

Key Documents

SAB3701447

Sigma-Aldrich

Monoclonal Anti-IDO2 antibody produced in mouse

clone 1HC, purified immunoglobulin

Synonym(s):

Anti-3-dioxygenase 2, Anti-Ido, Anti-Ido2, Anti-Indol1, Anti-Indoleamine 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 408.00

CHF 408.00


Available to ship on11 April 2025Details



Select a Size

Change View
100 μG
CHF 408.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 408.00


Available to ship on11 April 2025Details


biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1HC, monoclonal

form

buffered aqueous solution

species reactivity

mouse

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Ido2(209176)

Specificity

Anti-IDO2 was purified from concentrated tissue culture supernate by Protein G chromatography followed by extensive dialysis against the buffer stated above. IDO2 antibody is specific for mouse IDO2 protein. Cross-reactivity with IDO2 from other sources has not been determined.

Immunogen

Complete mouse IDO2 protein followed by hybridoma development.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied in 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% (w/v) Sodium Azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ivana Campia et al.
PloS one, 10(5), e0126159-e0126159 (2015-05-09)
Multidrug resistant cancer cells are hard to eradicate for the inefficacy of conventional anticancer drugs. Besides escaping the cytotoxic effects of chemotherapy, they also bypass the pro-immunogenic effects induced by anticancer drugs: indeed they are not well recognized by host
Alexander Lercher et al.
PLoS pathogens, 16(10), e1008973-e1008973 (2020-10-13)
The liver is a central regulator of metabolic homeostasis and serum metabolite levels. Hepatocytes are the functional units of the liver parenchyma and not only responsible for turnover of biomolecules but also act as central immune signaling platforms. Hepatotropic viruses

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service