Skip to Content
Merck
All Photos(1)

Key Documents

SAB3501181

Sigma-Aldrich

Anti-Bonzo antibody produced in rabbit

purified antibody (DEAE)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 377.00

CHF 377.00


Estimated to ship on01 May 2025



Select a Size

Change View
100 μG
CHF 377.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 377.00


Estimated to ship on01 May 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified antibody (DEAE)

antibody product type

primary antibodies

clone

polyclonal

form

liquid

species reactivity

human

concentration

1 mg/mL

technique(s)

western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CXCR6(10663)

General description

Bonzo Antibody: Human immunodeficiency virus (HIV) and simian immunodeficiency virus (SIV) require coreceptors, in addition to CD4, to infect target cells. Some G protein-coupled receptors including CCR5, CXCR4, CCR3, and CCR2b in the chemokine receptor family have been identified as HIV coreceptors. An orphan G protein-coupled receptor was recently cloned and designated Bonzo, STRL33 and TYMSTR, and identified as HIV and SIV coreceptor. Bonzo/STRL33 is used by SIV, HIV-2 and HIV-1. The messenger RNA of Bonzo/STRL33 is expressed in lymphoid tissues and activated peripheral blood lymphocytes.

Immunogen

Bonzo antibody was raised against a 20 amino acid peptide near the carboxy terminus of human Bonzo.The immunogen is located within the last 50 amino acids of Bonzo.

Application

Bonzo antibody can be used for detection of Bonzo by Western blot at 1:1000 dilution. Antibody validated: Western Blot in human samples. All other applications and species not yet tested.

Physical form

Supplied in PBS containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service